Få mere at vide
Loxiglumide, Tocris Bioscience™
CCK1 Receptor Antagonists
Brand: Tocris Bioscience 3036/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Loxiglumide is a CCK1 antagonist. Suppresses CCK-induced proliferation of acinar cells. Inhibits pancreatic secretion of digestive enzymes. Active in vivo.Tekniske data
| 107097-80-3 | |
| 4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid | |
| 5 mg |