Learn More
Kautex™ LDPE Wide Mouth Bottles without Closures
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
Brand:  Kautex™ 2000070529
 View more versions of this product
Product Code. 10129760
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Closures available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
LDPE
No
Round
Wide Neck
1Each
No
Wide Neck Bottle
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Kautex™ LDPE Wide Mouth Bottles without Closures > Capacity: 100mL