Learn More
Lovibond™ Ammonia Reagent Set Powder Packs™
Premeasured unit dose reagent for water quality testing
Brand: Lovibond™ 535500
Additional Details : Weight : 0.50000kg Transport : UN number : 2680 Chem class : 8 Pack group : II
Includes: Ammonia Cyanurate Reagent (10mL sample, 100 pack), Ammonia Salicylate Reagent (10mL sample, 100 pack)
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
10mL ammonia cyanurate reagent (100 pack), 10mL ammonia salicylate reagent (100 pack) | |
0 to 0.8mg/L | |
Ammonia Nitrogen Reagent Set |
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers | |
100 | |
Salicylate |