Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Manganese
For use with MC500 multi-parameter colorimeter
Brand:  Lovibond™ 517621BT
Additional Details : Weight : 0.30000kg Transport : UN number : 2680 Chem class : 8 Pack group : II
Product Code. 15311915
  • 1015.00 DKK / Pack of 100
Estimated Shipment: 25-06-2024
to see stock.
Includes:
  • Manganese LR No. 1 Tablets, 100 pack
  • Manganese LR No. 2 Tablets, 100 pack
Description
Description
  • Parameter: Manganese
  • Method: Formaldoxime
  • Sample Size: 10mL
Specifications
Specifications
Tablet
MC500 multi-parameter colorimeter
100
Formaldoxime
Water quality testing
Manganese No. 1 LR tablets (100 pack), manganese No. 2 LR tablets (100 pack)
0.02 to 4mg/L
Manganese Reagent Set