missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Aqua Comparitor Color Discs: Iron
For testing water with the Aqua Comparator
195.00 DKK
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
11775448
|
Lovibond™
146220 |
Iron HR Color Disc | 0.05 to 1mg/L |
195.00 DKK
Each |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
11775538
|
Lovibond™
146320 |
Iron HR Color Disc | 1 to 10mg/L |
195.00 DKK
Each |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for iron using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ |