Learn More
Lovastatin, 98%, BLD Pharm™
Product Code. 31448618
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31448618 100 mg
31438892 100 g
31448256 500 g
31442181 5 g
31436144 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31448618 Supplier BLD Pharm Supplier No. BD136706100mg
 Restricted Item
Forventet forsendelse: 25-11-2026
Specifications
CAS 75330-75-5
Molecular Formula C24H36O5
MDL Number MFCD00072164
Quantity 100 mg
SMILES CC[C@H](C)C(O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(O3)=O)[C@@]12[H])=O
Molecular Weight (g/mol) 404.54
Percent Purity 98%
Chemical Name or Material Lovastatin