Learn More
Lotoxane™, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Specifications
Color | Colorless |
Boiling Point | 180°C |
Packaging | Metal can |
Quantity | 25 L |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Lotoxane]TS
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P501-Dispose of contents/container to .