Få mere at vide
Losartan Potassium EP Impurity K, Mikromol™
Beskrivelse
Losartan Potassium EP Impurity K, Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Losartan Carboxaldehyde |
| CAS | 114798-36-6 |
| Analyt- eller komponentnavne | Losartan Carboxaldehyde |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H21 Cl N6 O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-carbaldehyde, EXP 3179, DUP 167, Losartan Potassium Imp. K (EP), Losartan Imp. K (EP), Losartan USP Related Compound C, Losartan USP RC C, 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-carbaldehyde, Losartan Carboxaldehyde, Losartan Imp K (EP), Losartan Related Compound C (USP), Losartan Related Compound C, Losartan Potassium Impurity K, Losartan Impurity K |
| InChI formel | InChI=1S/C22H21ClN6O/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-12,14H,2-3,8,13H2,1H3,(H,25,26,27,28) |
| Løsningstype | Neat |
| SMIL | CCCCc1nc(Cl)c(C=O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4 |
| Vis mere |