Få mere at vide
Losartan EP Impurity F, Mikromol™
Artikelnummer. 17984436
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17984436 25 mg
17930676 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17984436 Leverandør Mikromol Leverandørnr. MM0168.150025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Losartan EP Impurity F
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Losartan Isopropyl Ether
CAS 1332713-64-0
Analyt- eller komponentnavne Losartan Isopropyl Ether
Forsendelsestilstand Room Temperature
Molekylær formel C25 H29 Cl N6 O
Certificeringer/compliance ISO 17025
Synonym 5-[4'[[2-Butyl-4-chloro-5-[[(1-methylethyl)oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-1H-tetrazole (Losartan Isopropyl Ether), Losartan Potassium Imp. F (EP), Losartan Imp. F (EP), 5-[4'[[2-Butyl-4-chloro-5-[[(1-methylethyl)oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-1H-tetrazole, Losartan Isopropyl Ether, Losartan Potassium Impurity F, Losartan Impurity F
InChI formel InChI=1S/C25H29ClN6O/c1-4-5-10-23-27-24(26)22(16-33-17(2)3)32(23)15-18-11-13-19(14-12-18)20-8-6-7-9-21(20)25-28-30-31-29-25/h6-9,11-14,17H,4-5,10,15-16H2,1-3H3,(H,28,29,30,31)
SMIL CCCCc1nc(Cl)c(COC(C)C)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
IUPAC navn 5-[2-[4-[[2-butyl-4-chloro-5-(isopropoxymethyl)imidazol-1-yl]methyl]phenyl]phenyl]-1H-tetrazole
Molekylvægt (g/mol) 464.992
Mængde 25 mg
Formel vægt 464.2091 g/mol