Få mere at vide
Losartan Carboxaldehyde, TRC
Artikelnummer. 30482332
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482332 10 mg
30467657 100 mg
30474049 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482332 Leverandør Toronto Research Chemicals Leverandørnr. TRCL47050510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114798-36-6
Molekylær formel C22 H21 Cl N6 O
Molekylvægt (g/mol) 420.89
Synonym 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-carbaldehyde, Losartan Carboxaldehyde, Losartan Imp K (EP), Losartan Related Compound C (USP)
IUPAC navn 2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carbaldehyde
SMIL CCCCc1nc(Cl)c(C=O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
Tekniske data
Kemisk navn eller materiale Losartan Carboxaldehyde
Mængde 10 mg
InChI formel InChI=1S/C22H21ClN6O/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-12,14H,2-3,8,13H2,1H3,(H,25,26,27,28)
Formel vægt 420.1465