Få mere at vide
Losartan Azide, TRC
Artikelnummer. 30006946
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006946 50 mg
30007005 10 mg
30010926 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006946 Leverandør Toronto Research Chemicals Leverandørnr. TRCL47050150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 727718-93-6
Molekylær formel C22 H22 Cl N9
Molekylvægt (g/mol) 447.9234
Synonym 1H-Tetrazole, 5-[4'-[[5-(azidomethyl)-2-butyl-4-chloro-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]- (9CI), 5-[4'-[[5-(Azidomethyl)-2-butyl-4-chloro-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2H-tetrazole
IUPAC navn 5-[2-[4-[[5-(azidomethyl)-2-butyl-4-chloroimidazol-1-yl]methyl]phenyl]phenyl]-1H-tetrazole
SMIL CCCCc1nc(Cl)c(CN=[N+]=[N-])n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
Tekniske data
Kemisk navn eller materiale Losartan Azide
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C22H22ClN9/c1-2-3-8-20-26-21(23)19(13-25-29-24)32(20)14-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-27-30-31-28-22/h4-7,9-12H,2-3,8,13-14H2,1H3,(H,27,28,30,31)
Formel vægt 447.1687
Procent renhed >95%