missing translation for 'onlineSavingsMsg'
Få mere at vide
Loreclezole, MedChemExpress Artikelnummer.: 16466737

Loreclezole, MedChemExpress

Artikelnummer. p-200100849
5 mg
Encompass_Preferred
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16466737

Brand: Medchem Express HY105272

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

MedChemExpress Loreclezole, an antiepileptic compound, is a selective GABAA receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors.

Loreclezole, an antiepileptic compound, is a selective GABAA receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors.

Specifications

Kemisk navn eller materiale Loreclezole
Fysisk form Solid
CAS 117857-45-1
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C10H6Cl3N3
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym R 72063
Opløselighedsinformation DMSO : 110 mg/mL (400.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL Cl/C(C1=C(C=C(Cl)C=C1)Cl)=C\N2C=NC=N2
Molekylvægt (g/mol) 274.53
Formel vægt 274.53
Procent renhed 99.68%
Grad Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Loreclezole, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.