Få mere at vide
Tocris Bioscience™ Loreclezole hydrochloride
GREENER_CHOICE
Artikelnummer. 30001230
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001230 50 mg
30001229 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001230 Leverandør Tocris Bioscience™ Leverandørnr. 1295/50
Subtype-selective GABAA modulator
Subtype-selective GABAA receptor modulator. Acts as a positive allosteric modulator of β2 or β3-subunit containing receptors. Also acts as a negative modulator at a novel regulatory site, enhancing GABAA receptor sensitization. Inhibits homomeric ρ1 GABAC receptors.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2227372-56-5
Molekylær formel C10H7Cl4N3
Tekniske data
Kemisk navn eller materiale Loreclezole hydrochloride
Mængde 50 mg
Formel vægt 311
Renhed >99%
Molekylvægt (g/mol) 310.987