Få mere at vide
Loratadine-d5 (ethyl-d5), TRC
Artikelnummer. 30468382
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30468382 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468382 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4695815MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Loratadine-d5 (ethyl-d5)
CAS 1398065-63-8
Molekylær formel C22 D5 H18 Cl N2 O2
Mængde 5 mg
Synonym Ethyl-d5 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate, Loratadine-d5, Loratadine-D5 (ethyl-D5)
InChI formel InChI=1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
Molekylvægt (g/mol) 387.914
Formel vægt 387.176