Få mere at vide
Loratadine-d5 (ethyl-d5), CDN
Artikelnummer. 30308825
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308825 0.05 g
30315702 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308825 Leverandør CDN Leverandørnr. CDND73920.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Loratadine-d5 (ethyl-d5)
CAS 1398065-63-8
Molekylær formel C22 D5 H18 Cl N2 O2
Mængde 0.05 g
Synonym Ethyl-d5 4-(8-chloro-5,6-dihydro-11 H-benzo[5,6 ]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate, Loratadine-d5, Loratadine-D5 (ethyl-D5)
InChI formel InChI=1 S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14 H,2,5-6,9-10,12-13H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])OC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 4-(8-chloro-5,6-dihydrobenzo[1,2 ]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
Molekylvægt (g/mol) 387.914
Formel vægt 387.176 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity