Få mere at vide
Loratadine-d4, TRC
Artikelnummer. 30477744
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30477744 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477744 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46957710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Loratadine-d4
CAS 381727-27-1
Renhedsgrad noter HPLC
Molekylær formel C22H19D4ClN2O2
Mængde 10 mg
Synonym 4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylic Acid-d5, Ethyl Ester
InChI formel InChI=1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3/i12D2,13D2
SMIL ClC1=CC(CCC2=C/3N=CC=C2)=C(C=C1)C3=C4CC([2H])([2H])N(C(OCC)=O)C([2H])([2H])C\4
Molekylvægt (g/mol) 386.91
Procent renhed >95