Få mere at vide
Loratadine-d4 Epoxide N-Oxide, TRC
Artikelnummer. 30466871
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30466871 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466871 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46959210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Loratadine-d4 Epoxide N-Oxide
CAS 1794754-14-5
Molekylær formel C22H19D4ClN2O4
Mængde 10 mg
InChI formel InChI=1S/C22H23ClN2O4/c1-2-28-20(26)24-12-9-21(10-13-24)22(29-21)18-8-7-17(23)14-16(18)6-5-15-4-3-11-25(27)19(15)22/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3/i12D2,13D2
SMIL [2H]C1([2H])CC2(CC([2H])([2H])N1C(=O)OCC)OC23c4ccc(Cl)cc4CCc5cccn(=O)c35
Molekylvægt (g/mol) 418.91
Formel vægt 418.1597