Learn More
Lopinavir, 98%, Thermo Scientific Chemicals Product Code.: 15315088
Lopinavir, 98%, Thermo Scientific Chemicals
Product Code. p-7084039 Shop All Thermo Scientific Chemicals Products
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Product Code. 15315088
Brand: Thermo Scientific Acros 456750010
Request Bulk or Custom Format
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Lopinavir, CAS # 192725-17-0, is an antiretroviral drug of the amphetamine class.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Lopinavir is a retroviral protease inhibitor that acts as an antiretroviral and can prevent the propagation of human immunodeficiency virus (HIV) and hepatitis viruses within cells
• Lopinavir is a dicarboxylic acid diamide and a substituted form of amphetamine
Applications
• Lopinavir may be used in the laboratory study of other retroviruses
• Lopinavir is suitable for laboratory research of drug delivery systems; some studies have shown that the activity of lopinavir can be enhanced by alternative delivery methods such as microencapsulation
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 192725-17-0
Molecular Formula C37H48N4O5
Molecular Weight (g/mol) 628.81
MDL Number MFCD22628840
InChI Key KJHKTHWMRKYKJE-SUGCFTRWSA-N
PubChem CID 133109001
IUPAC Name N-[(2R,4R,5R)-5-[[2-(2,6-dimethylphenoxy)acetyl]amino]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamideShow MoreShow Less
SMILES CC(C)[C@H](N1CCCNC1=O)C(=O)N[C@H](C[C@H](O)[C@H](CC1=CC=CC=C1)NC(=O)COC1=C(C)C=CC=C1C)CC1=CC=CC=C1
Specifications
Quantity 1 g
Chemical Name or Material Lopinavir
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0418444 Certificate TypeCertificate of Analysis Date22/04/2024 Product Code456750010,456750050
Lot NumberA0434453 Certificate TypeCertificate of Analysis Date18/10/2023 Product Code456750010,456750050
Lot NumberA0422665 Certificate TypeCertificate of Analysis Date25/09/2023 Product Code456750010,456750050
Lot NumberA0403710 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code456750010,456750050
Lot NumberA0414166 Certificate TypeCertificate of Analysis Date12/12/2022 Product Code456750010,456750050
Product Identifier
  • Lopinavir
  • Multiple Manufacturers
    Exclusive savings, limited-time offer.
    Promo Code: 24112
    Expires2025-06-27
    New Offer
    Save Now
RUO – Research Use Only
Product Content Correction
Product Title
Lopinavir, 98%, Thermo Scientific Chemicals >