Learn More
Lonza™ Reliant™ Precast Gels
For rapid and reproducible resolution of DNA sizes from 8bp-10kb - Lonza™ Reliant™ Precast Gels are ideal for analysis of PCR, restriction digest fragments or most cloning needs.
Brand: Lonza™ 54825
Additional Details : Weight : 1.00000kg
Alert:
Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.
Description
- Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
- Easy to handle
- Fit most common mini submerged electrophoresis systems
- Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
Specifications
TBE | |
2% | |
Precast Gel | |
12-well | |
2% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide | |
Agarose | |
Yes | |
0.19 in. | |
≤15 μL | |
9.5 cm | |
6 cm |
Clear | |
Multiwell | |
100 to ≥3000 bp | |
Yes | |
DNA separation by gel electrophoresis | |
20 Gels | |
18° to 26°C | |
0.5 cm | |
3.7 in. | |
2.37 in. |