missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ ProSieve™ Color Protein Markers

Visible markers for use in SDS-PAGE gels

Brand:  Lonza™ 193837

Additional Details : CAS Number : 151-21-3 Weight : 0.90720kg

 View more versions of this product

Product Code. 11518005

  • 2868.00 DKK / 500µL


Estimated Shipment
02-06-2021
Add to basket

Description

Description

  • Useful for monitoring the efficiency of separation during electrophoresis
  • Confirm that transfer has occurred from gel to membrane in Western blotting

ProSieve Color Protein Marker

  • Proteins in the marker have been labeled with 3 different fluorescent dyes
  • Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load.
  • Proteins have approximate masses of 10, 15, 20, 25, 40, 50, 80, 125, and 190kDa
  • Each lot is individually calibrated against a ProSieve Unstained Protein Marker
    
ProSieve QuadColor Protein Marker
  • Proteins are individually prestained using four different dyes, producing a brightly colored ladder with an easy-to-remember pattern
  • Marker is supplied ready-to-use, no heating, further dilution or addition of a reducing agent is required before use
  • Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250, and 300kDa
  • Apparent molecular weights are provided for each lot

Specifications

Specifications

Protein Molecular Weight Marker/Ladder
Liquid
−20° to 4°C
500 μL
Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting
Prestained using four different dyes, Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250 and 300kd
4.6 to 300 kd
Videos
SDS
Documents