Learn More
Lonza™ Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions
A selection of concentrated and premix solutions for automated, high accuracy DNA sequencing applications. Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions offer greater flexibility to optimize run conditions and change gel percentages.
Brand: Lonza™ 50615
Additional Details : Weight : 0.45000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III
Description
Concentrated Solutions- Offer more flexibility to optimize run conditions and change gel percentages
- Require the addition of urea, buffer, APS, and TEMED
- Contain all gel components for increased run-to-run consistency and ease of use
- Require the addition of APS and TEMED for gel polymerization
Specifications
Agarose Electrophoresis Gels | |
DNA and RNA Sequencing, genotyping | |
1 L |
Liquid | |
Agarose | |
18° to 26°C |