missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ PowerCHO™ CD Medium
Support the growth of CHO K1, DXB11, DG44 and GS systems
1125.00 DKK
Specifications
Content And Storage | 2° to 8°C |
---|---|
Cell Line | CHO |
Without Additives | No Glutamine, No Phenol Red, No Hypoxanthine, No Thymidine |
Quantity | 1 L |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11615260
|
Lonza™
BE12-771Q |
PowerCHO 2; 1L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
PowerCHO 1, 2, and 3 Chemically Defined, Serum-free CHO Media are the next generation in CHO media, optimized for both cell growth and protein production. For therapeutic bioprocessing applications, these protein-free formulations also facilitate both downstream purification and regulatory compliance.
- Non-animal origin media
- Hydrolysate-free, serum-free
- Contains HEPES and Pluronic™ F-68
- Free of glutamine, hypoxanthine, thymidine and phenol red
- Maintain high viability (>90%) at high cell densities
- Support high-density, large scale production volumes through balanced formulation
Specifications
2° to 8°C | |
CHO | |
No Glutamine, No Phenol Red, No Hypoxanthine, No Thymidine | |
1 L |