Learn More
Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium
Used to provide buffer system to maintain cell culture media in the physiological pH range of 7 to 7.6.
Brand:  Lonza™
Additional Details : Weight : 1.36080kg
 View more versions of this product
Product Code. 10110035
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
  • Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
Specifications
Specifications
15° to 30°C
No Calcium, No Magnesium
500 mL
Dulbecco's Phosphate Buffered Saline
7 to 7.6