missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ CHO (Chinese Hamster Ovary) Serum-Free Culture Media: ProCHO5

For optimized expression of protein in CHO cells

Brand:  Lonza™ BE12-766Q

Additional Details : Weight : 0.10000kg

Product Code. 12764119

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • ProCHO-5 supports increased protein production of CHO cells already growing in suspension

†Store as detailed on label.

Specifications

Specifications

2° to 8°C
CHO Medium
No Hypoxanthine, No Thymidine, No Glutamine
Liquid
Phenol Red, 0.1% Pluronic F-68
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers