Learn More
Lonza™ AccuGENE™ PBS
Ready to use solutions ideal for a wide range of molecular biology applications
Brand: Lonza™ 51225
Additional Details : Weight : 1.00000kg
Description
AccuGENE Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
- High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
- Efficient - Ready-made solutions eliminate experiment preparation time.
- Flexible - Customized solutions are available to meet individual needs
Specifications
18° to 26°C | |
1000mL | |
Ideal for a wide range of molecular biology applications |
1 X PBS | |
KH2PO4 1.7mM, Na2HPO4 5mM, NaCl 150mM | |
7.4 |