Få mere at vide
Tocris Bioscience™ Lomeguatrib
GREENER_CHOICE
Artikelnummer. 18094662
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
18094662 10 mg
1 options
Artikelnummer. 18094662 Leverandør Tocris Bioscience™ Leverandørnr. 4359/10
MGMT inhibitor
Lomeguatrib (LomG) is a potent inhibitor of O6-methylguanine-DNA methyltransferase (MGMT); (IC50 = 0.009 μM in cell-free extracts from HeLa S3 cells); LomG attenuates MGMT activity in vitro and in vivo. LomG enhances the antitumor activity of temozolomide (Cat. No. 2706) in both human melanoma and MCF-7 xenografts. LomG acts as a pseudosubstrate for MGMT and can be used to induce rapid ubiquitin proteasome pathway degradation of proteins that have a MGMT fusion.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 192441-08-0
Molekylær formel C10H8BrN5OS
Tekniske data
Kemisk navn eller materiale Lomeguatrib
Mængde 10 mg
Formel vægt 326.17
Renhed >98%
Molekylvægt (g/mol) 326.17