Få mere at vide
Loganin, TRC
Artikelnummer. 30471343
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471343 100 mg
30475931 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471343 Leverandør Toronto Research Chemicals Leverandørnr. TRCL469405100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18524-94-2
Molekylær formel C17 H26 O10
Molekylvægt (g/mol) 390.38
Synonym (1S,4aS,6S,7R,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-6-hydroxy-7-methylcyclopenta[c]pyran-4-carboxylic Acid Methyl Ester, (-)-Loganin, Loganoside, (1S)-1α-(β-D-Glucopyranosyloxy)-1,4aα,5,6,7,7aα-hexahydro-6α-hydroxy-7α-methylcyclopenta[c]pyran-4-carboxylic Acid Methyl Ester, 7-Hydroxy-6-desoxyverbenalin, NSC 606403
IUPAC navn methyl (1S,4aS,6S,7R,7aS)-6-hydroxy-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate
SMIL COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]3[C@@H](C)[C@@H](O)C[C@H]13
Tekniske data
Kemisk navn eller materiale Loganin
Mængde 100 mg
InChI formel InChI=1S/C17H26O10/c1-6-9(19)3-7-8(15(23)24-2)5-25-16(11(6)7)27-17-14(22)13(21)12(20)10(4-18)26-17/h5-7,9-14,16-22H,3-4H2,1-2H3/t6-,7+,9-,10+,11+,12+,13-,14+,16-,17-/m0/s1
Formel vægt 390.1526