Få mere at vide
Lofepramine-d3, TRC
Artikelnummer. 30466718
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30466718 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466718 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4693821MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lofepramine-d3
CAS 1185083-78-6
Molekylær formel C26H24D3ClN2O
Mængde 1 mg
Synonym 1-(4-Chlorophenyl)-2-[[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl](methyl-d3)amino]ethanone, 4'-Chloro-2-[[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl- (methyl-d3)-amino]acetophenone, Amplit-d3, Leo 640-d3, Lopramine-d3
InChI formel InChI=1S/C26H27ClN2O/c1-28(19-26(30)22-13-15-23(27)16-14-22)17-6-18-29-24-9-4-2-7-20(24)11-12-21-8-3-5-10-25(21)29/h2-5,7-10,13-16H,6,11-12,17-19H2,1H3/i1D3
SMIL ClC(C=C1)=CC=C1C(CN(C([2H])([2H])[2H])CCCN2C(C=CC=C3)=C3CCC4=CC=CC=C42)=O
Molekylvægt (g/mol) 421.98