missing translation for 'onlineSavingsMsg'
Learn More
Lodenafil, MedChemExpress Product Code.: 16495088

Lodenafil, MedChemExpress

Product Code. p-200101912
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495088

Brand: Medchem Express HY123210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lodenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor for the treatment of erectile dysfunction (ED).

Lodenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor for the treatment of erectile dysfunction (ED).

Specifications

Chemical Name or Material Lodenafil
CAS 139755-85-4
Color White
Purity Grade Notes Research
Molecular Formula C23H32N6O5S
Quantity 10 mg
Synonym Hydroxyhomosildenafil
Solubility Information DMSO : ≥ 250 mg/mL (495.44 mM)
SMILES O=C1C2=C(NC(C3=CC(S(=O)(N4CCN(CC4)CCO)=O)=CC=C3OCC)=N1)C(CCC)=NN2C
Molecular Weight (g/mol) 504.6
Formula Weight 504.6
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Lodenafil, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.