missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOC388813 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06030AF405
Additional Details : Weight : 0.00970kg
Description
LOC388813 Polyclonal specifically detects LOC388813 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LOC388813 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
LOC388813 uncharacterized protein ENSP00000383407-like | |
Produced in rabbits immunized with E. coli-derived Human LOC388813/ AF165138.7 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
388813 | |
Store at 4°C in the dark. | |
IgG |