Få mere at vide
Tocris Bioscience™ LM22B 10
Beskrivelse
LM22B 10 is a TrkB and TrkC agonist. Exhibits neurotrophic activity (EC50 = 200-300 nM), improves survival and increases neurite outgrowth in hippocampal cells in vitro. Exhibits no significant activity in a screen of 57 other receptors and transporters. Inhibits binding of BDNF and neurotrophin-3 to TrkB and TrkC, respectively. Active in vivo and brain penetrant.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2,2′,2′,2′′′-[[(4-Chlorophenyl)methylene]bis(4,1-phenylenenitrilo)]tetrakisethanol |
| CAS | 342777-54-2 |
| Mængde | 10 mg |
| Molekylær formel | C27H33ClN2O4 |
| Renhed | 98% |