Få mere at vide
Lithocholic-2,2,4,4-d4 Acid, CDN
Artikelnummer. 30313785
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313785 0.1 g 0.1g
30315716 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313785 Leverandør CDN Leverandørnr. CDND37420.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lithocholic acid-2,2,4,4-d4
CAS 83701-16-0
Molekylær formel C24 2H4 H36 O3
Mængde 0.1 g
Synonym Cholan-24-oic-2,2,4,4-d4 acid, 3-hydroxy-, (3α,5β)-, (3α,5β)-3-Hydroxycholan-24-oic-2,2,4,4-d4 acid, Lithocholic acid-d4, Lithocholic Acid-2,2,4,4-d4
InChI formel InChI=1 S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25 H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1/i10D2,14D2
SMIL [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 CC[C@@H]2 C([2 H])([2 H])[C@@H]1 O)[C@H](C)CCC(=O)O
IUPAC navn (4 R)-4-[(3 R,5 R,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-3-hydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 380.6
Formel vægt 380.3229 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity