Få mere at vide
Lithium Tetrafluoroborate, Sigma-Aldrich™
Beskrivelse
We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. LITHIUM TETRAFLUOROBORATE is a key component in developing greener energy solutions, particularly in lithium-ion batteries. Its unique properties enhance ionic conductivity and stability, making it an effective electrolyte that improves battery performance. By utilizing this eco-friendly compound, we can advance sustainable energy technologies while reducing reliance on less environmentally friendly materials.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Lithium Tetrafluoroborate |
| Smeltepunkt | 324°C (Decomposition) |
| CAS | 14283-07-9 |
| Mængde | 10 g |
| MDL nummer | MFCD00011087 |
| Synonym | Lithium tetrafluoroborate, Lithium tetrafluoroborate |
| Molekylvægt (g/mol) | 93.75 |
| Grad | Research |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.