Learn More
Lithium for AAS 1g/L HNO3 matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical B030500
Additional Details : Weight : 0.50000kg
Quantity | 500mL |
---|
Chemical Identifiers
Specifications
1g/L | |
HNO3 | |
500mL |