Learn More
Lithium nitrate, anhydrous, 99.999% (metals basis)
Description
Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon

Specifications
Specifications
Boiling Point | 600°C (decomposition) |
Melting Point | 264°C |
Color | White |
Physical Form | Crystalline/Lump |
Quantity | 10 g |
UN Number | UN2722 |
Merck Index | 14,5536 |
Solubility Information | Soluble in water,ethanol,methanol,pyridine,ammonia and acetone. |
Formula Weight | 68.95 |
Percent Purity | 99.999% |
Show More |
Safety and Handling


- Lithium nitrate, anhydrous
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- MIXTURE LIST-Contains : Lithium nitrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.