missing translation for 'onlineSavingsMsg'
Få mere at vide

Lithium niobium oxide, 99.99%

Artikelnummer. 11463398
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
50 g
250 g
Pakningsstørrelse:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11463398 250 g 250g
11418850 5 g 5g
11453398 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11463398 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A17805.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 12031-63-9 | LiNbO3 | 147.84 g/mol

It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 12031-63-9
Molekylær formel LiNbO3
Molekylvægt (g/mol) 147.84
MDL nummer MFCD00011080
InChI nøgle GQYHUHYESMUTHG-UHFFFAOYSA-N
Synonym Lithium niobate
PubChem CID 23665650
SMIL [Li+].[O-][Nb](=O)=O

Tekniske data

Kemisk navn eller materiale Lithium niobium oxide
Smeltepunkt ∼1275°C
Mængde 250 g
Opløselighedsinformation Soluble in water.
IUPAC navn lithium(1+) niobiumoylolate
Formel vægt 147.85
Procent renhed 99.99%
Lugt Odorless
Tæthed 4.3

RUO– Kun brug til forskning

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.