Få mere at vide
Lithium niobium oxide, 99.99%
Description
It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
Specifications
Specifications
| Kemisk navn eller materiale | Lithium niobium oxide |
| Smeltepunkt | ∼1275°C |
| Mængde | 5 g |
| Opløselighedsinformation | Soluble in water. |
| IUPAC navn | lithium;oxido(dioxo)niobium |
| Formel vægt | 147.85 |
| Procent renhed | 99.99% |
| Lugt | Odorless |
| Tæthed | 4.3 |
RUO– Kun brug til forskning
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.