missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide, ultra dry, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407575
Click to view available options
Quantity:
10 g
50 g
250 g
10377-51-2
ILi
133.84
MFCD00011092
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
66321
lithium;iodide
[Li+].[I-]
10377-51-2
ILi
133.84
MFCD00011092
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
66321
lithium;iodide
[Li+].[I-]

Light Sensitive and Hygroscopic

It is used as a absorbent in refrigeration process, as a catalyst, and in photography. It is also useful for organic synthesis, pessat iodination, cleavage of esters, ethers, epoxides, decarboethoxylation, carbonylation, and aldol condensation. It is used for manufacturing medicines and mineral waters. It is used in electochemicstry and redox reactions. Lithium iodide is also a catalyzed conjugate addition reaction of β-dicarbonyl compounds. It is also useful for tetrathioquinodimethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1170°C to 1190°C
Melting Point 449°C
Color White
Physical Form Beads
Quantity 10 g
Assay Percent Range 99% (Metal basis)
Solubility Information Soluble in water,alcohol,acetone,methanol and ammonium hydoxide.
Formula Weight 133.84
Odor Odorless
Refractive Index 1.955
Sensitivity Hygroscopic; Light sensitive
Appearance White powder
Density 4.076 g/cm3 at 20°C
Chemical Name or Material Lithium iodide, ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'