missing translation for 'onlineSavingsMsg'
Learn More
Lithium hydroxide monohydrate, 99.995% (metals basis) Product Code.: 11470800

Lithium hydroxide monohydrate, 99.995% (metals basis)

Product Code. p-4781972
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11470800

Brand: Thermo Scientific Alfa Aesar 044473.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lithium hydroxide monohydrate, H3LiO2, CAS Number-1310-66-3, unii-g51xlp968g, lithium hydroxide monohydrate, lioh water, lithiumhydrate, hydroxyde de lithium, lithium hydroxido, lithium hydroxide, monohydrate, lithium hydroxide hydrate, lioh.hydrate, lioh-hydrate, 100g, 99.995% (Metals basis), White | CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 41.96 (23.95 Anhydrous)
Density 1.51 g/cm3 at 20°C
Chemical Name or Material Lithium hydroxide monohydrate
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Lithium hydroxide monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium hydroxide monohydrate, 99.995% (metals basis) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.