missing translation for 'onlineSavingsMsg'
Learn More

Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals

Catalog Number p-7050716
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
16733-97-4
C5H5Li
72.04
MFCD00001439,MFCD00001439
CSOHRTAOCDVTQU-UHFFFAOYSA-N
lithium cyclopentadienide, cyclopentadienyllithium, acmc-1bo1f, lithium cyclopenta-1,3-dien-1-ide, lithium 1+ ion cyclopenta-1,3-dien-1-ide
14063469
lithium;cyclopenta-1,3-diene
[Li+].C1C=CC=[C-]1
This item is not returnable. View return policy
16733-97-4
C5H5Li
72.04
MFCD00001439,MFCD00001439
CSOHRTAOCDVTQU-UHFFFAOYSA-N
lithium cyclopentadienide, cyclopentadienyllithium, acmc-1bo1f, lithium cyclopenta-1,3-dien-1-ide, lithium 1+ ion cyclopenta-1,3-dien-1-ide
14063469
lithium;cyclopenta-1,3-diene
[Li+].C1C=CC=[C-]1

Lithium cyclopentadienide is a useful reagent in preparing metallocenes and cyclopentadienyl derivatives. It is also used as intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN3180
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 72.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lithium cyclopentadienide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'