missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lithium chloride, 99%, for molecular biology

Catalog Number p-4813515
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1382.0°C
Melting Point 605.0°C
Quantity 100 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiCl
Merck Index 15,5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
Formula Weight 42.38
Percent Purity 99%
Grade Molecular Biology
Packaging Plastic bottle
Specific Gravity 2.06
Density 2.0600g/mL
Chemical Name or Material Lithium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Lithium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Lithium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'