Learn More
Thermo Scientific™ Listeria Selective Agar (Oxford Formulation)
Isolate and detect Listeria monocytogenes from clinical, food and environmental samples with this selective, diagnostic, ready-to-use medium.
Brand: Thermo Scientific™ PO5026A
118.63 DKK valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Additional Details : Weight : 0.40000kg
Description
Isolate and detect Listeria monocytogenes from clinical, food and environmental samples with the diagnostic Thermo Scientific™ Oxoid™ Listeria Selective Agar (Oxford Formulation). The medium is based on the formulations described by Curtis et al.1 Listeria monocytogenes, being capable of hydrolyzing aesculin, produce black zones around the colonies, while most other unwanted Gram-positive flora is inhibited. This medium is the specified plating medium in the FDA/BAM isolation procedure2 and in standardised testing methods of other national and international bodies3.
Foodborne infection by Listeria monocytogenes has prompted increased concern for detecting this organism in foods, in the environment and in pathological specimens from both human and animal subjects. The ability to isolate the organism has been impeded in the past by lack of an effective selective medium, as Listeria monocytogenes can be easily and completely overgrown by competing flora.
Use Listeria Selective Agar (Oxford Formulation) for selective isolation and detection of Listeria monocytogenes from clinical and food specimens.
- Easy-to-read: Listeria monocytogenes form brown colored colonies with black zones due to aesculin hydrolysis.
- An FDA/BAM specified plating medium for Listeria monocytogenes isolation3.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Lemon Yellow | |
Listeria Selective Agar (Oxford Formulation) | |
Germany | |
19.5g±1.0.g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
10 x 90mm plates, film wrapped |