Få mere at vide
Lipoic acid, MedChemExpress
Artikelnummer. 16471896
Encompass_Preferred
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
16471896 500 mg
1 options
Artikelnummer. 16471896 Leverandør Medchem Express Leverandørnr. HY18733
MedChemExpress Lipoic acid ((R)-(+)-α-Lipoic acid) is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. (R)-(+)-α-Lipoic acid is more effective than racemic Lipoic acid.
Lipoic acid ((R)-(+)-α-Lipoic acid) is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. (R)-(+)-α-Lipoic acid is more effective than racemic Lipoic acid.
Tekniske data
Kemisk navn eller materiale Lipoic acid
Fysisk form Solid
CAS 1200-22-2
Farve Pale Yellow
Mængde 500 mg
Renhedsgrad noter Research
Molekylær formel C8H14O2S2
Til brug med (applikation) COVID-19-immunoregulation
Synonym (R)-(+)-α-Lipoic acid R-(+)-Thioctic acid
Opløselighedsinformation DMSO : ≥ 100 mg/mL (484.66 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(O)CCCC[C@H]1SSCC1
Molekylvægt (g/mol) 206.33
Formel vægt 206.33
Procent renhed 98.4%