Learn More
Linseed Oil, Pure, Raw, ≈d= 0.93
Brand: Fisher Chemical L/1950/17
Additional Details : CAS Number : 8001-26-1 Weight : 2.50000kg
Quantity | 2.5L |
---|
Chemical Identifiers
8001-26-1 | |
131507 |
Specifications
Amber glass bottle | |
Yellow | |
2.5L | |
131507 |
315.6°C | |
-23.9°C | |
8001-26-1 | |
Liquid |
Safety and Handling
- Linseed oil
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.