missing translation for 'onlineSavingsMsg'
Få mere at vide

Linoleoyl Carnitine, TRC

Artikelnummer. 30466004
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466004 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30466004 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46820050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Linoleoyl Carnitine
CAS 36816-10-1
Renhedsgrad noter HPLC
Molekylær formel C25 H45 N O4
Mængde 50 mg
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z,12Z)-1-oxo-9,12-octadecadien-1-yl]oxy]-, inner salt, (2R)-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxo-9,12-octadecadienyl)oxy]-, inner salt, [R-(Z,Z)]-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z,12Z)-1-oxo-9,12-octadecadienyl]oxy]-, inner salt, (2R)- (9CI), L-Linoleoylcarnitine, Linoleoylcarnitine, Linoleylcarnitine
InChI formel InChI=1S/C25H45NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4/h9-10,12-13,23H,5-8,11,14-22H2,1-4H3/b10-9-,13-12-/t23-/m1/s1
SMIL CCCCC\C=C/C\C=C/CCCCCCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C
IUPAC navn (3R)-3-[(9Z,12Z)-octadeca-9,12-dienoyl]oxy-4-(trimethylazaniumyl)butanoate
Molekylvægt (g/mol) 423.63
Formel vægt 423.3349
Procent renhed >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.