Få mere at vide
Lindane-d6, TRC
Artikelnummer. 30472136
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30472136 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472136 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4659915MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale gamma-HCH D6
CAS 60556-82-3
Molekylær formel C6 2H6 Cl6
Mængde 5 mg
Synonym (1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane-1,2,3,4,5,6-d6, Hexadeuterated lindane, Lindane-d6, gamma-HCH D6
InChI formel InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+/i1D,2D,3D,4D,5D,6D
SMIL [2H][C@]1(Cl)[C@@]([2H])(Cl)[C@]([2H])(Cl)[C@@]([2H])(Cl)[C@@]([2H])(Cl)[C@]1([2H])Cl
IUPAC navn 1,2,3,4,5,6-hexachloro-1,2,3,4,5,6-hexadeuteriocyclohexane
Molekylvægt (g/mol) 296.87
Formel vægt 293.8977