Få mere at vide
Lincomycin 2-Palmitate Hydrochloride, TRC
Artikelnummer. 30479918
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479918 5 mg
30471201 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479918 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4663005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23295-14-9
Molekylær formel C34H65ClN2O7S
Molekylvægt (g/mol) 681.41
Synonym trans-α-Methyl 6,8-Dideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-D-erythro-D-galacto-octopyranoside 2-Palmitate Monohydrochloride, (2S-trans)-Methyl 6,8-dideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-D-erythro-α-D-galacto-octopyranoside 2-Hexadecanoate Monohydrochloride
IUPAC navn 4,5-dihydroxy-6-(2-hydroxy-1-(1-methyl-4-propylpyrrolidine-2-carboxamido)propyl)-2-(methylthio)tetrahydro-2H-pyran-3-yl palmitate hydrochloride
SMIL O=C(CCCCCCCCCCCCCCC)OC1C(SC)OC(C(C(C)O)NC(C2N(CC(C2)CCC)C)=O)C(C1O)O.Cl
Tekniske data
Kemisk navn eller materiale Lincomycin 2-Palmitate Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C34H64N2O7S.ClH/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-21-27(38)42-32-30(40)29(39)31(43-34(32)44-5)28(24(3)37)35-33(41)26-22-25(20-7-2)23-36(26)4;/h24-26,28-32,34,37,39-40H,6-23H2,1-5H3,(H,35,41);1H
Formel vægt 680.42