Få mere at vide
(±)-Linalool-d3 (vinyl-d3), CDN
Artikelnummer. 30319618
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319618 0.01 g
30324670 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319618 Leverandør CDN Leverandørnr. CDND69610.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Linalool-d3 (vinyl-d3)
CAS 1216673-02-7
Molekylær formel C10 D3 H15 O
Mængde 0.01 g
InChI formel InChI=1 S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11 H,1,6,8H2,2-4H3/i1D2,5 D
SMIL [2 H]C(=C([2 H])C(C)(O)CCC=C(C)C)[2 H]
IUPAC navn 1,1,2-trideuterio-3,7-dimethylocta-1,6-dien-3-ol
Molekylvægt (g/mol) 157.27
Formel vægt 157.1546 g/mol
Procent renhed 99 atom % D, min 96% Chemical Purity