Få mere at vide
Linagliptin-d3, TRC
Artikelnummer. 30478775
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30478775 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478775 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46590210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Linagliptin D3 (2-butyn-1-yl-4,4,4 D3)
CAS 1398044-48-8
Molekylær formel C25 D3 H25 N8 O2
Mængde 10 mg
Synonym 1H-Purine-2,6-dione, 8-[(3R)-3-amino-1-piperidinyl]-7-(2-butyn-1-yl-4,4,4-d3)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-, 8-[(3R)-3-Amino-1-piperidinyl]-7-(2-butyn-1-yl-4,4,4-d3)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-p
InChI formel InChI=1S/C25H28N8O2/c1-4-5-13-32-21-22(29-24(32)31-12-8-9-17(26)14-31)30(3)25(35)33(23(21)34)15-20-27-16(2)18-10-6-7-11-19(18)28-20/h6-7,10-11,17H,8-9,12-15,26H2,1-3H3/t17-/m1/s1/i1D3
SMIL [2H]C([2H])([2H])C#CCn1c(nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12)N5CCC[C@@H](N)C5
IUPAC navn 8-[(3R)-3-aminopiperidin-1-yl]-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]-7-(4,4,4-trideuteriobut-2-ynyl)purine-2,6-dione
Molekylvægt (g/mol) 475.561
Formel vægt 475.252