Få mere at vide
Linadryl Hydrochloride, LoGiCal
Artikelnummer. 30458642
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30458642 5 mg
1 options
Artikelnummer. 30458642 Leverandør LoGiCal Leverandørnr. LGCFOR3072.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Linadryl Hydrochloride
CAS 5928-69-8
Analyt- eller komponentnavne Linadryl Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H23 N O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym 4-[2-(Diphenylmethoxy)ethyl]morpholine hydrochloride (1:1), 4-[2-(Benzhydryloxy)ethyl]morpholine hydrochloride, Linadryl hydrochloride
InChI formel InChI=1S/C19H23NO2.ClH/c1-3-7-17(8-4-1)19(18-9-5-2-6-10-18)22-16-13-20-11-14-21-15-12-20;/h1-10,19H,11-16H2;1H
Løsningstype Neat
SMIL Cl.C(CN1CCOCC1)OC(c2ccccc2)c3ccccc3
IUPAC navn 4-(2-benzhydryloxyethyl)morpholine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 333.85
Mængde 5 mg
Formel vægt 333.1496 g/mol